Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
MAP7D1 (MAP7 Domain Containing 1) is a Protein Coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: arginine/proline rich coiled-coil 1; Arginine/proline-rich coiled-coil domain-containing protein 1; FLJ10350; FLJ39022; MAP7 domain-containing protein 1; proline arginine rich coiled coil 1; Proline/arginine-rich coiled-coil domain-containing protein 1
Gene Aliases: KIAA1187; MAP7D1; PARCC1; PP2464; RPRC1
UniProt ID: (Human) Q3KQU3
Entrez Gene ID: (Human) 55700
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support