Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSGPRAGFYR QELNKTVWEV PQRLQGLRPV GSGAYGSVCS AYDARLRQKV AVKKLSRPFQ SLIHARRTYR ELRLLKHLKH ENVIGLLDVF TPATSIEDFS EVYLVTTLMG ADLNNIVKCQ ALSDEHVQFL VYQLLRGLKY IHSAGIIHRD LKPSNVAVNE DCELRILDFG LARQADEEMT GYVATRWYRA PEIMLNWMHY NQTVDIWSVG CIMAELLQGK ALFPGSDYID QLKRIMEVVG TPSPEVLAKI SSEHARTYIQ SLPPMPQKDL SSIFRGANPL AIDLLGRMLV LDSDQRVSAA EALAHAYFSQ YHDPEDEPEA EPYDESVEAK ERTLEEWKEL TYQEVLSFKP PEPPKPPGSL EIEQ
MAPK11 (p38 beta) is a serine/threonine kinase which is involved in a variety of cellular processes, and is activated by proinflammatory cytokines and environmental stress. Mitogen-activated protein kinase 11. The protein encoded by p38 MAPK beta is a member of the MAP kinase family. MAP kinases act as an integration point for multiple biochemical signals, and are involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation, and development. This kinase is most closely related to p38 MAP kinase, both of which can be activated by proinflammatory cytokines and environmental stress. This kinase is activated through its phosphorylation by MAP kinase kinases (MKKs), preferably by MKK6. Transcription factor ATF2/CREB2 has been shown to be a substrate of this kinase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MAP kinase 11; MAP kinase p38 beta; Mitogen-activated protein kinase 11; Mitogen-activated protein kinase p38 beta; mitogen-activated protein kinase p38-2; OTTHUMP00000196655; p38-2; SAPK2b; Stress-activated protein kinase 2b; stress-activated protein kinase-2; stress-activated protein kinase-2b
Gene Aliases: MAPK11; p38-2; P38B; p38Beta; P38BETA2; PRKM11; SAPK2; SAPK2B
UniProt ID: (Human) Q15759
Entrez Gene ID: (Human) 5600
Molecular Function: non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support