Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: ACRKVAVKKI ALSDARSMKH ALREIKIIRR LDHDNIVKVY EVLGPKGTDL QGELFKFSVA YIVQEYMETD LARLLEQGTL AEEHAKLFMY QLLRGLKYIH SANVLHRDLK
Mitogen-activated protein kinase 4 is a member of the mitogen-activated protein kinase family. Tyrosine kinase growth factor receptors activate mitogen-activated protein kinases which then translocate into the nucleus where it phosphorylates nuclear targets.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ERK-4; Erk3-related; Extracellular signal-regulated kinase 4; MAP kinase 4; MAP kinase isoform p63; MAPK 4; Mitogen-activated protein kinase 4; p63-MAPK
Gene Aliases: ERK-4; ERK4; MAPK4; p63-MAPK; p63MAPK; PRKM4
UniProt ID: (Human) P31152
Entrez Gene ID: (Human) 5596
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support