Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The protein encoded by this gene is a regulator of the pancreatic beta-cell function. It is highly similar to JIP-1, a mouse protein known to be a regulator of c-Jun amino-terminal kinase (Mapk8). This protein has been shown to prevent MAPK8 mediated activation of transcription factors, and decrease IL-1 beta and MAP kinase kinase 1 (MEKK1) induced apoptosis in pancreatic beta cells. This protein also functions as a DNA-binding transactivator of the glucose transporter GLUT2. RE1-silencing transcription factor (REST) is reported to repress the expression of this gene in insulin-secreting beta cells. This gene is found to be mutated in a type 2 diabetes family, and thus is thought to be a susceptibility gene for type 2 diabetes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C-Jun-amino-terminal kinase-interacting protein 1; IB-1; Islet-brain 1; Islet-brain-1; JIP-1; JIP-1-related protein; JNK MAP kinase scaffold protein 1; JNK-interacting protein 1; JRP; mitogen activated protein kinase 8 interacting protein; mitogen activated protein kinase 8 interacting protein 1; Mitogen-activated protein kinase 8-interacting protein 1; PRKM8 interacting protein; protein kinase, mitogen-activated 8 interacting protein
Gene Aliases: IB1; JIP-1; JIP1; Mapk8ip; MAPK8IP1; mjip-2a; PRKM8IP; Skip
UniProt ID: (Human) Q9UQF2, (Rat) Q9R237, (Mouse) Q9WVI9
Entrez Gene ID: (Human) 9479, (Rat) 116457, (Mouse) 19099
Molecular Function: scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support