Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
                        
                        This antibody is predicted to react with bovine and human based on sequence homology.
This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.
MARCH10 is a member of the MARCH family of membrane-bound E3 ubiquitin ligases to target lysines in substrate proteins, thereby signaling their vesicular transport between membrane compartments.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 10-Mar; FLJ35757; MARCH-X; membrane associated ring finger 10; membrane-associated ring finger (C3HC4) 10, E3 ubiquitin protein ligase; Membrane-associated RING finger protein 10; Membrane-associated RING-CH protein X; microtubule-associated E3 ubiquitin ligase; probable E3 ubiquitin-protein ligase MARCH10; Probable E3 ubiquitin-protein ligase MARCHF10; RING finger protein 190; RING-type E3 ubiquitin transferase MARCHF10; testis secretory sperm-binding protein Li 228n
Gene Aliases: MARCH-X; MARCH10; MARCHF10; RGD1311692; RNF190
UniProt ID: (Human) Q8NA82, (Rat) Q5XIV2
Entrez Gene ID: (Human) 162333, (Rat) 303596
				If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
				Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support