Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 From June 2-13 take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 3
Immunogen sequence: LNSIFPGIGC PVPRIPAEAN PLADHVSATR ILCGALVFPT IATIVGKLMF SSVNSNLQRT ILGGIAFVAI KGAFKVYFKQ QQYLRQAHRK ILNYPEQEEA
MARCH5 is a ubiquitin ligase of the mitochondrial outer membrane that plays a role in the control of mitochondrial morphology by regulating mitofusin-2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase MARCH5; E3 ubiquitin-protein ligase MARCHF5; MARCH-V; membrane associated ring finger 5; membrane-associated ring finger (C3HC4) 5; Membrane-associated RING finger protein 5; Membrane-associated RING-CH protein V; Mitochondrial ubiquitin ligase; MITOL; RING finger protein 153; RING-type E3 ubiquitin transferase MARCHF5
Gene Aliases: 1810015H18Rik; 2310008I22Rik; 2700055A20Rik; 5730499H23Rik; E130202O05Rik; MARCH-V; MARCH5; MARCHF5; Marchv; MITOL; RNF153
UniProt ID: (Human) Q9NX47, (Mouse) Q3KNM2
Entrez Gene ID: (Human) 54708, (Rat) 294079, (Mouse) 69104
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support