Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MARCH6 is an E3 ubiquitin ligase that localizes to endoplasmic reticulum membranes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 6-Mar; Doa10 homolog; E3 ubiquitin-protein ligase MARCH6; E3 ubiquitin-protein ligase MARCHF6; MARCH-VI; membrane associated ring finger 6; membrane-associated ring finger (C3HC4) 6, E3 ubiquitin protein ligase; Membrane-associated RING finger protein 6; membrane-associated RING-CH finger protein 6; Membrane-associated RING-CH protein VI; Protein TEB-4; RING finger protein 176; RING-type E3 ubiquitin transferase MARCHF6
Gene Aliases: 3830408G03; DOA10; F830029L24Rik; KIAA0597; MARCH-VI; MARCH6; MARCHF6; mKIAA0597; RNF176; TEB4
UniProt ID: (Human) O60337, (Mouse) Q8BJA0
Entrez Gene ID: (Human) 10299, (Mouse) 223455
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support