Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MKVYIYSSGS VEAQKLLFGH STEGDILELV DGHFDTKIGH KVESESYRKI ADSIGCSTNN ILFLTDVTRE ASAAEEADVH VAVVVRPGNA GLTDDEKTYY SLITSFSELY LPSST
Bifunctional enzyme that catalyzes the enolization of 2,3-diketo-5-methylthiopentyl-1-phosphate (DK-MTP-1-P) into the intermediate 2-hydroxy-3-keto-5-methylthiopentenyl-1-phosphate (HK-MTPenyl-1-P), which is then dephosphorylated to form the acireductone 1,2-dihydroxy-3-keto-5-methylthiopentene (DHK-MTPene).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2,3-diketo-5-methylthio-1-phosphopentane phosphatase; acireductone synthase; Enolase-phosphatase E1; ENOPH1; MASA homolog
Gene Aliases: E1; ENOPH1; MASA; MST145; MSTP145; mtnC
UniProt ID: (Human) Q9UHY7
Entrez Gene ID: (Human) 58478
Molecular Function: phosphatase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support