Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody detects endogenous protein at a molecular weight of 76 kDa.
Purity is >95% by SDS-PAGE.
Mannose (or mannan)-binding lectin (MBL), also known as serum mannosebinding protein (MBP), initiates the lectin branch of the innate immune response by binding to the surface of potentially pathogenic microorganisms and initiating complement fixation through an N-terminal collagen-like domain. MBL is a key component in immune response in that it can directly trigger neutralization of invading microorganisms by an Ab-independent mechanism. Mutations of human MBL are associated with immunodeficiency resulting from a reduction in the ability of the mutant MBL to initiate complement fixation. In human, three types of MBL-associated serine proteases, MASP-1, MASP-2 and MASP-3, and a truncated form of MASP-2 (small MBL-associated protein; sMAP or MAp19) complex with MBL to activate the lectin pathway of the complement system. Activated MASPs subsequently cleave and activate downstream components of the complement pathway. MASP-3 is an alternatively spliced product from the MASP-1 gene and may function to inhibit MASP-2 by competing for MBL binding and inhibiting the activation of MBLassociated MASP-2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: mannan-binding; mannan-binding lectin serine peptidase 1 pseudogene 1; mannan-binding lectin serine protease 1 pseudogene 1; Mannan-binding lectin serine protease 2; Mannose-binding protein-associated serine protease 2; MASP-2; MBL-associated plasma protein of 19 kD; MBL-associated serine protease 2; small MBL-associated protein
Gene Aliases: MAP19; MASP-2; MASP1P1; MASP2; sMAP
UniProt ID: (Human) O00187, (Mouse) Q91WP0
Entrez Gene ID: (Human) 10747, (Mouse) 17175
Molecular Function: serine protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support