Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive Samples: HepG2, 293T, Mouse liver, Rat liver
Immunogen sequence: MNGPVDGLCD HSLSEGVFMF TSESVGEGHP DKICDQISDA VLDAHLKQDP NAKVACETVC KTGMVLLCGE ITSMAMVDYQ RVVRDTIKHI GYDDSAKGFD FKTCNVLVAL EQQSPDIAQC VHLDRNEEDV GAGDQGLMFG YATDETEECM PLTIILAHKL NARMADLRRS GLLPWLRPDS KTQVTVQYMQ DNGAVIPVRI HTIVISVQHN EDITLEEMRR ALKEQVIRAV VPAKYLDEDT VYHLQPSGRF VIGGPQGDAG VTGRKIIVDT YGGWGAHGGG AFSGKDYTKV DRSAAYAARW VAKSLVKAGL CRRVLVQVSY AIGVAEPLSI SIFTYGTSQK TERELLDVVH KNFDLRPGVI VRDLDLKKPI YQKTACYGHF GRSEFPWEVP RKLVF
MAT1A catalyzes the formation of S-adenosylmethionine from methionine and ATP. Methionine adenosyltransferase deficiency is caused by recessive and dominant mutations, the latter identified in autosomal dominant persistant hypermethioninemia.This gne encodes methionine adenosyltransferase I (alpha isoform), which catalyzes the formation of S-adenosylmethionine from methionine and ATP. Methionine adenosyltransferase deficiency is caused by recessive and dominant mutations, the latter identified in autosomal dominant persistant hypermethioninemia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AdoMet synthase 1; adoMet synthetase 1; MAT 1; MAT-I/III; Methionine adenosyltransferase 1; methionine adenosyltransferase I, alpha; Methionine adenosyltransferase I/III; S - adenosylmethionine synthetase; S-adenosylmethionine synthase isoform type-1; S-adenosylmethionine synthetase isoform type-1
Gene Aliases: AdoMet; AI046368; Ams; AMS1; MAT; MAT1A; MATA1; SADE; SAMS; SAMS1; SAS
UniProt ID: (Human) Q00266, (Rat) P13444, (Mouse) Q91X83
Entrez Gene ID: (Human) 4143, (Rat) 25331, (Mouse) 11720
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support