Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 3
Store as concentrated solution.
Centrifuge briefly prior to opening vial.
S adenosylmethionine synthetase catalyzes the formation of S adenosylmethionine from methionine and ATP. In mammalian tissues, there are three distinct forms of AdoMet synthases designated as alpha, beta, and gamma. Alpha and beta are expressed only in adult liver, while gamma is widely distributed in extrahepatic tissues.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AdoMet synthase 1; AdoMet synthase 2; adoMet synthetase 1; adoMet synthetase 2; MAT 1; MAT 2; MAT-I/III; MAT-II; Methionine adenosyltransferase 1; Methionine adenosyltransferase 2; methionine adenosyltransferase I, alpha; Methionine adenosyltransferase I/III; Methionine adenosyltransferase II; methionine adenosyltransferase II, alpha; S-adenosylmethionine synthase isoform type-1; S-adenosylmethionine synthase isoform type-2; S-adenosylmethionine synthetase isoform type-1; S-adenosylmethionine synthetase isoform type-2; testicular tissue protein Li 121
Gene Aliases: AdoMet; AI046368; Ams; AMS1; AMS2; D630045P18Rik; MAT; MAT1A; MAT2A; MATA1; MATA2; MATII; SAMS; SAMS1; SAMS2
UniProt ID: (Human) Q00266, (Human) P31153, (Mouse) Q91X83, (Mouse) Q3THS6
Entrez Gene ID: (Human) 4143, (Human) 4144, (Mouse) 11720, (Mouse) 232087
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support