Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 6
Positive Samples: Jurkat, HepG2, MCF7; Cellular Location: Cytoplasm, Cytoplasmic granule, Nucleus
Immunogen sequence: MAVSVTPIRD TKWLTLEVCR EFQRGTCSRP DTECKFAHPS KSCQVENGRV IACFDSLKGR CSRENCKYLH PPPHLKTQLE INGRNNLIQQ KNMAMLAQQM QLANAMMPGA PLQPVPMFSV APSLATNASA AAFNPYLGPV SPSLVPAEIL PTAPMLVTGN PGVPVPAAAA AAAQKLMRTD RLEVCREYQR GNCNRGENDC RFAHPADSTM IDTNDNTVTV CMDYIKGRCS REKCKYFHPP AHLQAKIKAA QYQVNQAAAA QAAATAAAMT QSAVKSLKRP LEATFDLGIP QAVLPPLPKR PALEKTNGAT AVFNTGIFQY QQALANMQLQ QHTAFLPPVP MVHGATPATV SAATTSATSV PFAATATANQ IPIISAEHLT SH
EGFR (Epidermal growth factor receptor, HER1, ErbB1) is encoded by the EGFR gene located on chromosome 7 in humans. EGFR belongs to the HER/ERbB family of proteins that includes three other receptor tyrosine kinases, ERbB2, ERbB3, ERbB4. EGFR is a transmembrane receptor and binding of its cognate ligands such as EGF (Epidermal Growth Factor) and TGF alpha (Transforming Growth Factor alpha) to the extracellular domain leads to EGFR dimerization followed by autophosphorylation of the tyrosine residues in the cytoplasmic domain. Phosphorylation of EGFR at certain residues is also mediated by Src-non-receptor kinase. EGFR activation signals multiple downstream signaling cascades such as the Ras - ERK, PI3-K - Akt, Jak - STAT and PKC pathways that help in growth and proliferation of cells. Phosphorylation of EGFR at Y1086 specifically allows binding of the adaptor protein GRB2, leading to activation of the MAPK pathway. Upon receptor activation and signaling, EGFR is endocytosed and targeted for degradation or recycling. Mutations in the EGFR gene are associated with lung cancer and multiple alternatively spliced transcript variants encode different protein isoforms of EGFR have been found. Increased production or activation of EGFR has been associated with poor prognosis in a variety of tumors. Moreover, EGFR overexpression is observed in tumors of the head and neck, brain, bladder, stomach, breast, lung, endometrium, cervix, vulva, ovary, esophagus, stomach and in squamous cell carcinoma.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp686P06174; EXP35; EXP40; EXP42; muscleblind-like 1; Muscleblind-like protein 1; Triplet-expansion RNA-binding protein
Gene Aliases: EXP; KIAA0428; MBNL; MBNL1; mKIAA0428
UniProt ID: (Human) Q9NR56, (Rat) A0A8I6B1J2, (Mouse) Q9JKP5
Entrez Gene ID: (Human) 4154, (Rat) 282635, (Mouse) 56758
Molecular Function:
RNA splicing factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support