Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 10
MC1R, a Melanocortin Receptor, mediates the effects of melanocyte-stimulating hormone (MSH) and adrenocorticotropic hormone (ACTH). MSH and its receptor, MC1R, are key regulators of melanogenesis. They control the relative proportions of red pheomelanin and black photoprotective eumelanin in the skin by increasing the production of eumelanin. Non-functional, truncated forms of the receptor lead to lighter coat color in animals. In contrast, constitutively active receptors lead to darker color.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alpha melanocyte stimulating hormone receptor; extension recessive yellow; MC1 receptor; MC1-R; melanocortin 1 receptor; melanocortin 1 receptor (alpha melanocyte stimulating hormone receptor); Melanocortin receptor 1; melanocortin-1 receptor; Melanocyte-stimulating hormone receptor; melanotropin receptor; MSH-R; tobacco darkening
Gene Aliases: CMM5; e; MC1R; MSH-R; MSHR; Mshra; SHEP2; Tob
UniProt ID: (Human) Q01726, (Mouse) Q01727
Entrez Gene ID: (Human) 4157, (Rat) 102552838, (Mouse) 17199
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support