Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button
            Target, gene symbol, antigen
            Application
            Target Species
            • Order Status
            • Quick Order
            • Promos
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • CD146 Antibodies

            NeoBiotechnologies

            MCAM (Melanoma Cell Adhesion Molecule)/MUC18/CD146 Monoclonal Antibody (C146/634)

            View all (101) CD146 antibodies

            Promotions

            View available promotion(s)

            Promo Code: RGLOW25 Save up to $900 and get the limited-edition holiday sweatshirt + matching beanie It’s been a year – this is your badge of honor for fluorescing through it all. Offer available until December 27.. Learn more

            Spend Now, Save Big Later! Spend Now, Save Big Later! Make your New year's resolution to save: earn up to $3,500 off a future January offer when you purchase a qualifying amount before December 31st. Learn more

            Datasheet
            Protocols
            Questions & Answers
            Catalog # 4162-MSM3-P0
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (USD)
            226.50
            Special offer
            Online exclusive
            Ends: 13-Dec-2025
            302.00
            Save 75.50 (25%)
             
            Check your price
            Request bulk or custom format
            Join Aspire™ for free antibodies
            Datasheet
            Protocols
            Questions & Answers

            Cite MCAM (Melanoma Cell Adhesion Molecule)/MUC18/CD146 Monoclonal Antibody (C146/634)

            • Antibody Testing Data (2)
            Application
            MCAM (Melanoma Cell Adhesion Molecule)/MUC18/CD146 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            Group 53 Created with Sketch.

            FIGURE: 1 / 2

            MCAM (Melanoma Cell Adhesion Molecule)/MUC18/CD146 Antibody (4162-MSM3-P0) in IHC (P)

            Formalin-fixed, paraffin-embedded human Melanoma stained with CD146 Monoclonal Antibody (C146/634).
            View Product

            MCAM (Melanoma Cell Adhesion Molecule)/MUC18/CD146 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            MCAM (Melanoma Cell Adhesion Molecule)/MUC18/CD146 Antibody in Immunohistochemistry (Paraffin) (IHC (P))

            Product Details

            4162-MSM3-P0

            Applications
            Tested Dilution
            Publications

            Immunohistochemistry (Paraffin) (IHC (P))

            Assay-dependent
            -

            Immunohistochemistry (PFA fixed) (IHC (PFA))

            1-2 µg/mL
            -

            Immunocytochemistry (ICC/IF)

            1-2 µg/mL
            -

            Flow Cytometry (Flow)

            1-2 µg/10^6 cells
            -
            Product Specifications

            Species Reactivity

            Human

            Host/Isotype

            Mouse / IgG1, kappa

            Class

            Monoclonal

            Type

            Antibody

            Clone

            C146/634

            Immunogen

            Recombinant human CD146 protein

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            200 µg/mL

            Purification

            Protein A/G

            Storage buffer

            PBS, pH 7.4, with 0.05% BSA

            Contains

            0.05% sodium azide

            Storage conditions

            4°C, do not freeze

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            Target Information

            CD146, also known as MCAM (Melanoma Cell Adhesion Molecule), is an integral membrane glycoprotein belonging to the immunoglobulin superfamily. It is heavily glycosylated, with more than 50% of its mass derived from carbohydrates. CD146 is primarily expressed on endothelial cells at cell-cell junctions, playing a crucial role in cell adhesion and the cohesion of the endothelial monolayer in vascular tissue. In addition to endothelial cells, CD146 is expressed on bone marrow fibroblasts, mesenchymal stromal cells, endometrial stromal cells, and some tumors, particularly melanoma. It has also been observed on a subset of circulating T cells and activated lymphocytes, but not on circulating endothelial cells, as indicated by the absence of other endothelial markers such as CD31 and CD51/61. CD146 functions as a calcium-independent cell adhesion molecule and may also act as a signal transduction molecule. It is involved in the recruitment of Fyn kinase and the subsequent tyrosine phosphorylation of intracellular proteins, which are important for actin cytoskeleton assembly. This signaling role includes triggering tyrosine phosphorylation of FYN and PTK2 and causing a transient increase in intracellular calcium concentration. In the context of cancer, CD146 expression may facilitate the interaction of melanoma cells with the vascular system, enhancing hematogenous tumor spread. Additionally, CD146 could serve as an adhesion molecule active in neural crest cells during embryonic development, highlighting its diverse roles in both normal physiological processes and disease states.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            References (0)

            Have you cited this product in a publication?
            Let us know so we can reference it here.
            Cite this product

            Bioinformatics

            Protein Aliases: CD146; Cell surface glycoprotein MUC18; Cell surface glycoprotein P1H12; Gicerin; melanoma adhesion molecule; Melanoma cell adhesion molecule; Melanoma-associated antigen A32; Melanoma-associated antigen MUC18; S-endo 1 endothelial-associated antigen

            View more View less

            Gene Aliases: CD146; MCAM; MUC18

            View more View less

            UniProt ID: (Human) P43121

            View more View less

            Entrez Gene ID: (Human) 4162

            View more View less

            Process(es)

            angiogenesis glomerular filtration cell adhesion anatomical structure morphogenesis positive regulation of cell migration vascular wound healing
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us About Us
            • Careers Careers
            • Investors Investors
            • News News
            • Social Responsibility Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
            • Policies and Notices
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-blue-555ccb4fbd-wx6js:80/100.66.76.49:80.
            git-commit: 75f0c7d4cb44686da2af39b21f2e6446a20282e2
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.41.0-2025.12.99-1.0