Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MARVLKAAA ANAVGLFSRL QAPIPTVRAS STSQPLDQVT GSVWNLGRLN HVAIAVPDLE KAAAFYKNIL GAQVSEAVPL PEHGVSVVFV NLGNTKMELL HPLGLDSPIA GFLQKNKAGG MHHICIEVDN INAAVMDLKK KKIRSLSEEV KIGAHGKPVI FLHPKDCGGV LVELEQA (1-176 aa encoded by BC020825)
MCEE catalyzes the interconversion of D- and L-methylmalonyl-CoA during the degradation of branched chain amino acids. odd chain-length fatty acids, and other metabolites. Mutations in this gene result in methylmalonyl-CoA epimerase deficiency, which is presented as mild to moderate methylmalonic aciduria.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DL-methylmalonyl-CoA racemase; glyoxalase domain containing 2; methylmalonyl CoA epimerase; Methylmalonyl-CoA epimerase, mitochondrial
Gene Aliases: 1110007A04Rik; GLOD2; MCEE
UniProt ID: (Human) Q96PE7, (Mouse) Q9D1I5
Entrez Gene ID: (Human) 84693, (Mouse) 73724, (Rat) 293829
Molecular Function: epimerase/racemase isomerase metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support