Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSVGAMKKG VGRAVGLGGG SGCQATEEDP LPDCGACAPG QGGRRWRLPQ PAWVEGSSAR LWEQATGTGW MDLEASLLPT GPNASNTSDG PDNLTSAGSP PRTGSISYIN IIMPSVFGTI CLLGIIGNST VIFAVVKKSK LHWCNNVPDI FIINLSVVDL LFLLGMPFMI HQLMGNGVWH FGETMCTLIT AMDANSQFTS TYILTAMAID RYLATVHPIS STKFRKPSVA TLVICLLWAL SFISITPVWL YARLIPFPGG AVGCGIRLPN PDTDLYWFTL YQFFLAFALP FVVITAAYVR ILQRMTSSVA PASQRSIRLR TKRVTRTAIA ICLVFFVCWA PYYVLQLTQL SISRPTLTFV YLYNAAISLG YANSCLNPFV YIVLCETFRK RLVLSVKPAA QGQLRAVSNA QTADEERTES KGT (1-422 aa encoded by BC021146)
MCHR1 is a member of the G protein-coupled receptor family 1. It is an integral plasma membrane protein which binds melanin-concentrating hormone. The encoded protein can inhibit cAMP accumulation and stimulate intracellular calcium flux, and is probably involved in the neuronal regulation of food consumption. Although structurally similar to somatostatin receptors, this protein does not seem to bind somatostatin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G protein coupled receptor 24; G protein-coupled receptor 24; G-protein coupled receptor 24; GPCR24; Gpr24-9 protein coupled receptor; MCH 1 receptor; MCH receptor 1; MCH-1R; MCH-R1; MCH1; MCH1R; MCHR; MCHR-1; Melanin-concentrating hormone receptor 1; SLC-1; Somatostatin receptor-like protein
Gene Aliases: AW049955; GPR24; MCH-1R; MCH1R; MCHR1; SLC-1; SLC1
UniProt ID: (Human) Q99705, (Mouse) Q8JZL2, (Rat) A0A140TAD3
Entrez Gene ID: (Human) 2847, (Mouse) 207911, (Rat) 83567
Molecular Function:
G-protein coupled receptor
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support