Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 17
Antibody detects endogenous levels of total MCRS1.
Modulates the transcription repressor activity of DAXX by recruiting it to the nucleolus (PubMed:11948183). As part of the NSL complex it may be involved in acetylation of nucleosomal histone H4 on several lysine residues (PubMed:20018852). Putative regulatory component of the chromatin remodeling INO80 complex which is involved in transcriptional regulation, DNA replication and probably DNA repair. May also be an inhibitor of TERT telomerase activity (PubMed:15044100). Binds to G-quadruplex structures in mRNA (PubMed:16571602). Binds to RNA homopolymer poly(G) and poly(U) (PubMed:16571602). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 58 kDa microspherule protein; cell cycle-regulated factor (78 kDa); Cell cycle-regulated factor p78; INO80 complex subunit J; INO80 complex subunit Q; MCRS2; Microspherule protein 1; nucleolar protein
Gene Aliases: C78274; ICP22BP; INO80Q; MCRS1; MCRS2; MSP58; P78
UniProt ID: (Human) Q96EZ8, (Mouse) Q99L90
Entrez Gene ID: (Human) 10445, (Rat) 300222, (Mouse) 51812
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support