Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LDIEVWGKD NKKHEERLGT CKVDISALPL KQANCLELPL DSCLGALLML VTLTPCAGVS VSDLCVCPLA DLSERKQITQ RYCLQNSLKD VKDVGILQVK VLKAADLLAA DFSGKSDPFC LLELGNDRLQ THTVYKNLNP EWNKVFTFPI KDIHDVLEVT VFDEDGDKPP DFLGKVAIPL LSIRDGQPNC YVLKNKDLEQ AFKGVIYLEM DLIYNPVKAS IRTFTPREKR FVEDSRKLSK KILSRDVDRV KRITMAIWNT MQFLKSCFQW ESTLRSTIAF AHRKEEPPLS MKCTLFSGDI GPSL (4-306 aa encoded by BC025708)
Might play a role in the development of cardiac outflow tract. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Multiple C2 and transmembrane domain-containing protein 2; multiple C2 domains, transmembrane 2; multiple C2-domains with two transmembrane regions 2 1
Gene Aliases: Gm489; MCTP2; RGD1562967
UniProt ID: (Human) Q6DN12, (Mouse) Q5RJH2
Entrez Gene ID: (Human) 55784, (Rat) 308742, (Mouse) 244049
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support