Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 企業服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Gene Expression
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins and Peptides
  • ELISA Kits
  • SNP Genotyping
  • 所有文檔和支持
  • 證書
  • SDS(安全資料表
  • 產品常見問題
  • Documents & Support
  • 手冊和試驗方法
    Search button Close
            Target of Interest
            Application
            Target Species
            • 訂單查詢
            • 快速訂購
            • 登入
              登入
              還沒有帳戶嗎 ? 立即註冊
              • 帳戶
              • 訂單
              • Connect: Lab, Data, Apps
              • 客製化產品與項目
              • 儀器管理

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • MDA5 Antibodies

            Invitrogen

            MDA5 Polyclonal Antibody

            View all (26) MDA5 antibodies
            Datasheet
            Protocols
            Questions & Answers
            Tech Support
            Datasheet
            Protocols
            Questions & Answers
            Tech Support

            Cite MDA5 Polyclonal Antibody

            • Antibody Testing Data (3)
            Application
            MDA5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            Group 53 Created with Sketch.

            FIGURE: 1 / 3

            MDA5 Antibody (PA5-27848) in IHC (P)

            MDA5 Polyclonal Antibody detects MDA5 protein at cytoplasm on human breast carcinoma by immunohistochemical analysis. Sample: Paraffin-embedded human breast carcinoma. MDA5 Polyclonal Antibody (Product # PA5-27848) diluted at 1:500. Antigen Retrieval: EDTA based buffer, pH 8.0, 15 min.
            View Product

            MDA5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            MDA5 Antibody in Western Blot (WB)
            MDA5 Antibody in Western Blot (WB)

            Product Details

            PA5-27848

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1:500-1:3,000
            -

            Immunohistochemistry (Paraffin) (IHC (P))

            1:100-1:1,000
            -
            Product Specifications

            Species Reactivity

            Human

            Host/Isotype

            Rabbit / IgG

            Class

            Polyclonal

            Type

            Antibody

            Immunogen

            Recombinant protein encompassing a sequence within the center region of human MDA5. The exact sequence is proprietary.

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            1 mg/mL

            Purification

            Antigen affinity chromatography

            Storage buffer

            PBS, pH 7, with 1% BSA, 20% glycerol

            Contains

            0.01% thimerosal

            Storage conditions

            Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            RRID

            AB_2545324

            Product Specific Information

            Recommended positive controls: THP-1, THP-1 (100 ng/ml PMA treatment for 16 hr and 1000 ng/ml LPS treatment for 24 hr).

            Predicted reactivity: Mouse (86%), Rat (86%), Pig (91%), Rhesus Monkey (98%).

            Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

            Target Information

            DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp and a protein kinase C-activating compound, mezerein. Irreversible reprogramming of melanomas can be achieved by treatment with both these agents; treatment with either agent alone only achieves reversible differentiation.

            ⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            References (0)

            Have you cited this product in a publication?
            Let us know so we can reference it here.
            Cite this product

            Bioinformatics

            Protein Aliases: CADM-140 autoantigen; Clinically amyopathic dermatomyositis autoantigen 140 kDa; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide; Helicard; Helicase with 2 CARD domains; interferon induced, with helicase C domain 1; Interferon-induced helicase C domain-containing protein 1; Interferon-induced with helicase C domain protein 1; MDA-5; melanoma differentiation associated protein-5; melanoma differentiation-associated gene 5; Melanoma differentiation-associated protein 5; MGC133047; Murabutide down-regulated protein; RIG-I-like receptor 2; RLR-2; RNA helicase-DEAD box protein 116

            View more View less

            Gene Aliases: AGS7; Hlcd; IDDM19; IFIH1; MDA-5; MDA5; RH116; RLR-2; SGMRT1

            View more View less

            UniProt ID: (Human) Q9BYX4

            View more View less

            Entrez Gene ID: (Human) 64135

            View more View less

            Function(s)

            DNA binding double-stranded RNA binding single-stranded RNA binding helicase activity protein binding ATP binding zinc ion binding ribonucleoprotein complex binding

            Process(es)

            detection of virus response to virus protein sumoylation negative regulation of type I interferon production positive regulation of interferon-alpha production positive regulation of interferon-beta production regulation of type III interferon production cytoplasmic pattern recognition receptor signaling pathway in response to virus MDA-5 signaling pathway innate immune response
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            產品訂購 Plus Icon Minus Icon
            • 訂單狀態
            • 訂購協助
            • 快速訂購
            • 現貨供應中心
            • 電子採購
            支持 Plus Icon Minus Icon
            • 協助與支持
            • 聯絡我們
            • 技術支持中心
            • 搜尋文件和證書
            • 回報網站問題
            資源 Plus Icon Minus Icon
            • 學習中心
            • 促銷活動
            • 活動和網路研討會
            • 社群媒體
            關於 Thermo Fisher Plus Icon Minus Icon
            • 關於我們 關於我們
            • 人才招募 人才招募
            • 投資者資訊 投資者資訊
            • 新聞 新聞
            • 企業社會責任 企業社會責任
            • 商標
            品牌 Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Policy
            • Price & Freight Policy
            • Cookie偏好

              在此網站使用 Cookie 的選擇

              我們以及我們的附屬公司和供應商使用 cookies 和類似技術來經營我們的網站、識別我們網站的訪客、提供安全登入、收集統計資料以優化網站功能,並提供根據您的興趣量身定制的內容。點擊「全部接受」 以接受所有 cookies,並直接進入網站,或點擊「管理設定」 以查看 cookies 類型的詳細說明,並選擇是否在網站接受某些 cookies。
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            Taiwan flag icon
            Taiwan

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-l9gpq:80/100.66.77.183:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0