Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
FIGURE: 1 / 1
AA Sequence of recombinant protein: YGANMEDSVC CRDYVRYRLP LRVVKHFYWT SDSCPRPGVV LLTFRDKEIC ADPRVPWVKM ILNKLSQ.
Preparation: Produced in BALB/c x ICR F1 mice immunized with highly pure Recombinant Human MDC (CCL22). Anti-Human MDC (CCL22)-specific antibody was purified from cell culture by Protein G affinity chromatography.
CCL22 may play a role in the trafficking of activated/effector T-lymphocytes to inflammatory sites and other aspects of activated T-lymphocyte physiology. Also, it is a chemotactic for monocytes. dendritic cells and natural killer cells. CCL22 is a mild chemoattractant for primary activated T-lymphocytes and a potent chemoattractant for chronically activated T-lymphocytes but has no chemattractant activity for neutrophils, eosinophils, and resting T-lymphocytes. Binds to CCR4.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AW060611; C-C motif chemokine 22; CC chemokine STCP-1; chemokine (C-C motif) ligand 22; Macrophage-derived chemokine; MDC(1-69); MGC108943; MGC34554; RP24-69A20.1; small inducible cytokine A22; small inducible cytokine subfamily A (Cys-Cys), member 22; Small-inducible cytokine A22; stimulated T cell chemotactic protein 1; Stimulated T-cell chemotactic protein 1
Gene Aliases: A-152E5.1; ABCD-1; CCL22; DC/B-CK; MDC; SCYA22; STCP-1
UniProt ID: (Human) O00626
Entrez Gene ID: (Human) 6367
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support