Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
MDC1 participates in the early response to DNA damage. It is thought to be a member of the BRCT (BRCA1 C-terminus) super family of nuclear proteins. It contains an N-terminus forkhead-associated (FHA) motif, two C-terminus BRCT motifs and 13 internal repetitions of a 41 amino acid sequence.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DAAP-285E11.7; DKFZp781A0122; homologue to Drosophila photoreceptor protein calphotin; mediation of DNA damage checkpoint 1; mediator of DNA damage checkpoint 1; Mediator of DNA damage checkpoint protein 1; mediator of DNA-damage checkpoint 1; MGC166888; Nuclear factor with BRCT domains 1
Gene Aliases: 6820401C03; AA413496; KIAA0170; MDC1; mKIAA0170; NFBD1
UniProt ID: (Human) Q14676, (Chimpanzee) Q7YR40, (Mouse) Q5PSV9
Entrez Gene ID: (Human) 9656, (Chimpanzee) 471959, (Mouse) 240087
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support