Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Immunogen sequence: MSEPIRVLV TGAAGQIAYS LLYSIGNGSV FGKDQPIILV LLDITPMMGV LDGVLMELQD CALPLLKDVI ATDKEDVAFK DLDVAILVGS MPRREGMERK DLLKANVKIF KSQGAALDKY AKKSVKVIVV GNPANTNCLT ASKSAPSIPK ENFSCLTRLD HNRAKAQIAL KLGVTANDVK NVIIWGNHSS TQYPDVNHAK VKLQGKEVGV YEALKDDSWL KGEFVTTVQQ RGAAVIKARK LSSAMSAAKA ICDHVRDIWF GTPEGEFVSM GVISDGNSYG VPDDLLYSFP VVIKNKTWKF VEGLPINDFS REKMDLTAKE LTEEKESAFE FLSSA (1-334 aa encoded by BC001484)
MDH1 is localized to the cytoplasm and may play pivotal roles in the malate-aspartate shuttle that operates in the metabolic coordination between cytosol and mitochondria.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Aromatic alpha-keto acid reductase; Cytosolic malate dehydrogenase; diiodophenylpyruvate reductase; epididymis secretory protein Li 32; KAR; malate dehydrogenase 1, NAD (soluble); Malate dehydrogenase, cytoplasmic; malate dehydrogenase, soluble; soluble malate dehydrogenase
Gene Aliases: B230377B03Rik; D17921; HEL-S-32; MDH-s; MDH1; MDHA; MGC:1375; Mor-2; MOR2
UniProt ID: (Human) P40925, (Mouse) P14152
Entrez Gene ID: (Human) 4190, (Mouse) 17449
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support