Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Knockdown to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 14
Recommended positive controls: 293T, A431, HeLa, HepG2, mouse brain, rat lung.
Predicted reactivity: Mouse (97%), Rat (96%), Zebrafish (90%), Xenopus laevis (91%), Cat (95%), Pig (95%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Malate dehydrogenase catalyzes the reversible oxidation of malate to oxaloacetate, utilizing the NAD/NADH cofactor system in the citric acid cycle. The protein encoded by this gene is localized to the mitochondria and may play pivotal roles in the malate-aspartate shuttle that operates in the metabolic coordination between cytosol and mitochondria. Several transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Sep 2013]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: malate dehydrogenase 2, NAD (mitochondrial); Malate dehydrogenase, mitochondrial; mitochondrial malate dehydrogenase 2, NAD; testicular tissue protein Li 120
Gene Aliases: M-MDH; MDH; Mdh-2; MDH2; MGC:3559; Mor-1; MOR1
UniProt ID: (Human) P40926, (Rat) Q6GSM4, (Mouse) P08249
Entrez Gene ID: (Human) 4191, (Rat) 81829, (Mouse) 17448
Molecular Function:
dehydrogenase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support