Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MARLPKLAV FDLDYTLWPF WVDTHVDPPF HKSSDGTVRD RRGQDVRLYP EVPEVLKRLQ SLGVPGAAAS RTSEIEGANQ LLELFDLFRC YLHSHPEWNE SSNSKSRVRD ICEGPNWAFE VQP (1-122 aa encoded by BC046912)
Magnesium-dependent phosphatase which may act as a tyrosine phosphatase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: fructosamine-6-phosphatase; Magnesium-dependent phosphatase 1; MDP-1
Gene Aliases: 1810034K20Rik; AI035604; FN6PASE; MDP-1; MDP1; RGD1311147
UniProt ID: (Human) Q86V88, (Mouse) Q9D967
Entrez Gene ID: (Human) 145553, (Mouse) 67881, (Rat) 290230
Molecular Function: hydrolase phosphatase metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support