Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Tissue lysate).
Immunogen sequence: MEPEAPRRRH THQRGYLLTR NPHLNKDLAF TLEERQQLNI HGLLPPSFNS QEIQVLRVVK NFEHLNSDFD RYLLLMDLQD RNEKLFYRVL TSDIEKFMPI VYTPTVGLAC QQYSLVFRKP RGLFITIHDR GHIASVLNAW PEDVIKAIVV TDGERILGLG DLGCNGMGIP VGKLALYTAC GGMNPQECLP VILDVGTENE ELLKDPLYIG LRQRRVRGSE YDDFLDEFME AVSSKYGMNC LIQFEDFANV NAFRLLNKYR NQYCTFNDDI QGTASVAVAG LLAALRITKN KLSDQTILFQ GAGEAALGIA HLIVMALEKE GLPKEKAIKK IWLVDSKGLI VKGRASLTQE KEKFAHEHEE MKNLEAIVQE IKPTALIGVA AIGGAFSEQI LKDMAAFNER PIIFALSNPT SKAECSAEQC YKITKGRAIF ASGSPFDPVT LPNGQTLYPG QGNNSYVFPG VALGVVACGL RQITDNIFLT TAEVIAQQVS DKHLEEGRLY PPLNTIRDVS LKIAEKIVKD AYQEKTATVY PEPQNKEAFV RSQMYSTDYD QILPDCYSWP EEVQKIQTKV DQ
ME1 encodes a cytosolic, NADP-dependent enzyme that generates NADPH for fatty acid biosynthesis. The activity of this enzyme, the reversible oxidative decarboxylation of malate, links the glycolytic and citric acid cycles. The regulation of expression for this gene is complex. Increased expression can result from elevated levels of thyroid hormones or by higher proportions of carbohydrates in the diet.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: malate dehydrogenase; malate dehydrogenase (oxaloacetate-decarboxylating) (NADP(+)); Malic enzyme 1; malic enzyme 1, NADP(+)-dependent, cytosolic; malic enzyme 1, soluble; Malic enzyme, cytoplasmic; NADP-dependent malic enzyme; NADP-ME; pyruvic-malic carboxylase
Gene Aliases: HUMNDME; ME1; MES
UniProt ID: (Human) P48163
Entrez Gene ID: (Human) 4199
Molecular Function:
oxidoreductase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support