Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Sequence of this protein is as follows: MVAGMLGLRE EKSEDQDLQG LKDKPLKFKK VKKDKKEEKE GKHEPVQPSA HHSAEPAEAG KAETSEGSGS APAVPEASAS PKQRRSIIRD RGPMYDDPTL PEGWTRKLKQ RKSGRSAGKY DVYLINPQGK AFRSKVELIA YFEKVGDTSL DPNDFDFTVT GRGSPSRREQ KPPKKPKSPK APGTGRGRGR PKGSGTTRPK AATSEGVQVK RVLEKSPGKL LVKMPFQTSP GGKAEGGGAT TSTQVMVIKR PGRKRKAEAD PQAIPKKRGR KPGSVVAAAA AEAKKKAVKE SSIRSVQETV LPIKKRKTRE TVSIEVKEVV KPLLVSTLGE KSGKGLKTCK SPGRKSKESS PKGRSSSASS PPKKEHHHHH HHSESPKAPV PLLPPLPPPP PEPESSEDPT SPPEPQDLSS SVCKEEKMPR GGSLESDGCP KEPAKTQPAV ATAATAAEKY KHRGEGERKD IVSSSMPRPN REEPVDSRTP VTERVS
MECP2 belongs to a family of nuclear proteins (including MBD1, MBD2, MBD3 and MBD4) that have a methyl-CpG binding domain. MECP2 is capable of binding specifically to methylated DNA. DNA methylation is the major modification of eukaryotic genomes and plays an essential role in mammalian development. MECP2 can also repress transcription from methylated gene promoters. In contrast to other MBD family members, MECP2 is X-linked and subject to X inactivation. MECP2 is dispensible in stem cells, but is essential for embryonic development. MECP2 gene mutations are the cause of most cases of Rett syndrome, a progressive neurologic developmental disorder and one of the most common causes of mental retardation in females.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp686A24160; MeCp-2 protein; methyl CpG binding protein 2; Methyl-CpG-binding protein 2; testis tissue sperm-binding protein Li 41a
Gene Aliases: 1500041B07Rik; AUTSX3; D630021H01Rik; Mbd5; MECP2; MRX16; MRX79; MRXS13; MRXSL; PPMX; RS; RTS; RTT; WBP10
UniProt ID: (Human) P51608, (Mouse) Q9Z2D6
Entrez Gene ID: (Human) 4204, (Mouse) 17257
Molecular Function:
DNA metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support