Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: MGVTCVSQM PVAEGKSVQQ TVELLTRKLE MLGAEKQGTF CVDCETYHTA ASTLGSQGQT GKLMYVMHNS EYPLSCFALF ENGPCLIADT NFDVLMVKLK GFFQSAKASK IETRGTRYQY CDFLVKVGTV TMGPSARGIS VEVEYGPCVV ASDCWSLLLE FLQSFLGSHT PGAPAVFGNR HDAVYGPADT MVQYMELFNK IRKQQQVPVA GIR (1-212 aa encoded by BC040950)
MED20 is a subunit of the Mediator complex, a multiprotein coactivator of RNA transcription that interacts with DNA-bound transcriptional activators, RNA polymerase II, and general initiation factors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hTRFP; Mediator complex subunit 20; Mediator of RNA polymerase II transcription subunit 20; Trf (TATA binding protein-related factor)-proximal homolog; TRF-proximal protein homolog
Gene Aliases: MED20; PRO0213; TRFP
UniProt ID: (Human) Q9H944
Entrez Gene ID: (Human) 9477
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support