Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MAAVDIRDNL LGISWVDSSW IPILNSGSVL DYFSERSNPF YDRTCNNEVV KMQRLTLEHL NQMVGIEYIL LHAQEPILFI IRKQQRQSPA QVIPLADYYI IAGVIYQAPD LGSVINSRVL TAVHGIQSAF DEAMSYCRYH PSKGYWWHFK DHEEQDKVRP KAKRKEEPSS IFQRQRVDAL LLDLRQKFPP KFVQLKPGEK PVPVDQTKKE AEPIPETVKP EEKETTKNVQ QTVSAKGPPE KRMRLQ
MED6 is a component of the Mediator complex, a coactivator involved in the regulated transcription of nearly all RNA polymerase II-dependent genes. Mediator functions as a bridge to convey information from gene-specific regulatory proteins to the basal RNA polymerase II transcription machinery. And mediator is recruited to promoters by direct interactions with regulatory proteins and serves as a scaffold for the assembly of a functional preinitiation complex with RNA polymerase II and the general transcription factors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Activator-recruited cofactor 33 kDa component; ARC33; CTD-2540L5.5; hMed6; Mediator complex subunit 6; Mediator of RNA polymerase II transcription subunit 6; Renal carcinoma antigen NY-REN-28
Gene Aliases: ARC33; MED6; NY-REN-28
UniProt ID: (Human) O75586
Entrez Gene ID: (Human) 10001
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support