Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The myocyte enhancer factor-2 (MEF-2) family of transcription factors associated with co-repessors or co-activators to regulate development and function of T cells, neuronal cells and muscle cells. Four family members arise from alternatively spliced transcripts, termed MEF-2A, -2B, -2C and -2D. These members bind as homo- and heterodimers to the MEF-2 site in the promoter region of affected genes. Differential regulation in the expression of the four transcripts implies functional distinction for each during embryogenesis and development. The process of differentiation from mesodermal precursor cells to myoblasts has led to the discovery of a variety of tissue-specific factors that regulate muscle gene expression.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: LOC729991-MEF2B; MADS box transcription enhancer factor 2, polypeptide B (myocyte enhancer factor 2B); MEF2BNB-MEF2B; Myocyte-specific enhancer factor 2B; RSRFR2; Serum response factor-like protein 2
Gene Aliases: AI451606; MEF2B; RSRFR2; XMEF2
UniProt ID: (Human) Q02080
Entrez Gene ID: (Human) 100271849, (Mouse) 17259
Molecular Function: MADS box transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support