Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAKTPSDHLL STLEELVPYD FEKFKFKLQN TSVQKEHSRI PRSQIQRARP VKMATLLVTY YGEEYAVQLT LQVLRAINQR LLAEELHRAA IQEYSTQENG TDDSAASSSL
This gene encodes a protein, also known as pyrin or marenostrin, that is an important modulator of innate immunity. Mutations in this gene are associated with Mediterranean fever, a hereditary periodic fever syndrome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HGNC:6998; Marenostrin; MGC126560; MGC126586; Pyrin
Gene Aliases: FMF; MEF; MEFV; TRIM20
UniProt ID: (Human) O15553
Entrez Gene ID: (Human) 4210
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support