Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: NPPFKSTAR FTLNPNPTGV QNPHIERLRT HSIESSGKLK ISPEQHWDFT AEDLKDLGEI GRGAYGSVNK MVHKPSGQIM AVKRIRSTVD EKEQKQLLMD LDVVMRSSDC PYIVQFYGAL FREGDCWICM ELMSTSFDKF YKYVYSVLDD VIPEEILGKI TLATVKALNH LKENLKIIHR DIKPSNILLD RSGNIKLCDF GISGQLVDSI AKTRDAGCRP YMAPERIDPS ASRQGYDVRS DVWSLGITLY ELATGRFPYP KWNSVFDQLT QVVKGDPPQL SNSEEREFSP SFINFVNLCL TKDESKRPKY KELLKHPFIL MYEERAVEVA CYVCKILDQM PATPSSPMYV D (50-399 aa encoded by BC036032)
MAP2K4 (MEK4) serine/threonine kinase (mitogen-activated protein kinase 4) MEK4 or MKK4 is a dual specificity kinase of the STE7 family that phosphorylates and activates JNK1 and -2 as well as p38. MAP2K4 preferentially phosphorylates the tyrosine residue of JNKs. MAP2K4 mediates a variety of physiological and pathophysiological processes such as responses to cellular stresses and inflammatory cytokines. MAP2K4 is important for carcinogenesis; a loss-off-function mutation in MAP2K4 was found in lung and pancreatic tumors. Conversely, a pro-oncogenic role was also found for MAP2K4.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C-JUN N-terminal kinase kinase 1; Dual specificity mitogen-activated protein kinase kinase 4; JNK kinase 1; JNK-activated kinase 1; JNK-activating kinase 1; JNKK; JNKK 1; MAP kinase kinase 4; MAPK/ERK kinase 4; MAPKK 4; MEK 4; mitogen activated protein kinase kinase 4, variant 1; SAPK kinase 1; SAPK/ERK kinase 1; SAPK/Erk/kinase 1; SEK1; Stress-activated protein kinase kinase 1
Gene Aliases: JNKK; JNKK1; MAP2K4; MAPKK4; MEK4; MKK4; PRKMK4; SAPKK-1; SAPKK1; SEK1; SERK1; SKK1
UniProt ID: (Human) P45985, (Mouse) P47809
Entrez Gene ID: (Human) 6416, (Mouse) 26398
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support