Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSQSKGKKR NPGLKIPKEA FEQPQTSSTP PRDLDSKACI SIGNQNFEVK ADDLEPIMEL GRGAYGVVEK MRHVPSGQIM AVKRIRATVN SQEQKRLLMD LDISMRTVDC PFTVTFYGAL FREGDVWICM ELMDTSLDKF YKQVIDKGQT IPEDILGKIA VSIVKALEHL HSKLSVIHRD VKPSNVLINA LGQVKMCDFG ISGYLVDSVA KTIDAGCKPY MAPERINPEL NQKGYSVKSD IWSLGITMIE LAILRFPYDS WGTPFQQLKQ VVEEPSPQLP ADKFSAEFVD FTSQCLKKNS KERPTYPELM QHPFFTLHES KGTDVASFVK LILGD (1-334 aa encoded by BC012009)
MKK6 (MEK6, MAP2K6) is a member of the dual specificity protein kinase family, which functions as a mitogen-activated protein (MAP) kinase kinase. MAP kinases, also known as extracellular signal-regulated kinases (ERKs), act as an integration point for multiple biochemical signals. This protein phosphorylates and activates p38 MAP kinase in response to inflammatory cytokines or environmental stress. As an essential component of p38 MAP kinase mediated signal transduction pathway, this gene is involved in many cellular processes such as stress induced cell cycle arrest, transcription activation and apoptosis. Tissue specificity: Isoform 2 is only expressed in skeletal muscle. Isoform 1, on the other hand, is found in skeletal muscle, heart, and in lesser extent in liver or pancreas.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dual specificity mitogen-activated protein kinase kinase 6; MAP kinase kinase 6; MAPK/ERK kinase 6; MAPKK 3; MAPKK 6; MAPKK3; MEK 6; MEK3; mitogen activated protein kinase kinase 6; MKK3; PRKMK3; protein kinase, mitogen activated, kinase 6; protein kinase, mitogen-activated, kinase 6 (MAP kinase kinase 6); SAPK kinase 3; SAPKK-2; SAPKK2; SAPKK3; Stress-activated protein kinase kinase 3
Gene Aliases: MAP2K6; MAPKK6; MEK6; MKK6; PRKMK6; SAPKK-3; SAPKK3; SKK3
UniProt ID: (Human) P52564, (Mouse) P70236
Entrez Gene ID: (Human) 5608, (Mouse) 26399
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support