Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • MEKK4 Antibodies

            Invitrogen

            MEKK4 Polyclonal Antibody

            View all (18) MEKK4 antibodies
            Datasheet
            Protocols
            Questions & Answers
            Datasheet
            Protocols
            Questions & Answers

            Cite MEKK4 Polyclonal Antibody

            • Antibody Testing Data (1)
            Application
            MEKK4 Antibody in Western Blot (WB)
            Group 53 Created with Sketch.

            FIGURE: 1 / 1

            MEKK4 Antibody (PA5-75856) in WB

            Western blot analysis of MEKK4 in Lane 1: A549 whole cell lysate, Lane 2: HEK293T whole cell lysate, Lane 3: PC12 whole cell lysate, Lane 4: SP20 whole cell lysate, Lane 5: H9C2 whole cell lysate, Lane 6: Raw264.7 whole cell lysate. Samples were incubated with MEKK4 polyclonal antibody (Product # PA5-75856) at a dilution of 1:500.
            View Product

            MEKK4 Antibody in Western Blot (WB)

            Product Details

            PA5-75856

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1:500-1:1,000
            -
            Product Specifications

            Species Reactivity

            Human, Mouse, Rat

            Host/Isotype

            Rabbit / IgG

            Class

            Polyclonal

            Type

            Antibody

            Immunogen

            Synthetic peptide corresponding to residues in Human MEKK4.

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            1 mg/mL

            Purification

            Antigen affinity chromatography

            Storage buffer

            PBS, pH 7.2, with 50% glycerol

            Contains

            0.02% sodium azide

            Storage conditions

            Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            RRID

            AB_2719584

            Product Specific Information

            The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

            Target Information

            This gene encodes a TBP- (TATA box-binding protein) associated phosphoprotein that represses both basal and activated levels of transcription. The encoded protein is phosphorylated in vivo and this phosphorylation affects its interaction with TBP. This protein contains a histone fold motif at the amino terminus, a TBP-binding domain, and a glutamine- and alanine-rich region. The binding of DR1 repressor complexes to TBP-promoter complexes may establish a mechanism in which an altered DNA conformation, together with the formation of higher order complexes, inhibits the assembly of the preinitiation complex and controls the rate of RNA polymerase II transcription.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            References (0)

            Have you cited this product in a publication?
            Let us know so we can reference it here.
            Cite this product

            Bioinformatics

            Protein Aliases: dJ473J16.1 (mitogen-activated protein kinase kinase kinase 4); JNKK1; MAP three kinase 1; MAP/ERK kinase kinase 4; MAP2K4; MAPK/ERK kinase kinase 4; MEK kinase 4; mitogen activated protein kinase kinase kinase 4; Mitogen-activated protein kinase kinase kinase 4; predicted protein of HQ0412; PRKMK4; SAPK/ERK kinase 1; SERK1; SSK2/SSK22 MAP kinase kinase kinase, yeast, homolog of; T-associated sex reversal

            View more View less

            Gene Aliases: D17Rp17; D17Rp17e; KIAA0213; MAP3K4; MAPKKK4; Mek4b; MEKK 4; MEKK4; mKIAA0213; MTK1; PRO0412; RP17; Rp17a; Tas

            View more View less

            UniProt ID: (Human) Q9Y6R4, (Mouse) O08648

            View more View less

            Entrez Gene ID: (Human) 4216, (Mouse) 26407, (Rat) 308106

            View more View less

            Function(s)

            MAP kinase kinase kinase activity protein binding ATP binding metal ion binding nucleotide binding protein kinase activity protein serine/threonine kinase activity MAP kinase kinase activity kinase activity transferase activity transferase activity, transferring phosphorus-containing groups molecular_function non-receptor serine/threonine protein kinase

            Process(es)

            MAPK cascade activation of MAPKK activity placenta development response to UV-C regulation of gene expression male germ-line sex determination positive regulation of telomere maintenance via telomerase intracellular signal transduction positive regulation of JUN kinase activity positive regulation of telomerase activity chorionic trophoblast cell differentiation positive regulation of p38MAPK cascade positive regulation of telomere capping activation of MAPK activity protein phosphorylation phosphorylation determination of dorsal identity biological_process
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            United States flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-584499df9d-2994x:80/100.66.79.195:80.
            git-commit: 256cec658627a60c8e1196038bf5905a13cd45ba
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.33.0-2025.07.30-1.0