Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The antibody detects endogenous levels of total MEOX2 protein.
Mesodermal transcription factor that plays a key role in somitogenesis and is required for sclerotome development. Activates expression of CDKN1A and CDKN2A in endothelial cells, acting as a regulator of vascular cell proliferation. While it activates CDKN1A in a DNA-dependent manner, it activates CDKN2A in a DNA-independent manner. May have a regulatory role when quiescent vascular smooth muscle cells reenter the cell cycle.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Growth arrest-specific homeobox; Homeobox protein MOX-2; mesenchyme homeo box 2 (growth arrest-specific homeo box); Mesenchyme homeobox 2
Gene Aliases: AI528662; GAX; MEOX2; Mox-2; MOX2
UniProt ID: (Human) P50222, (Rat) P39020, (Mouse) P32443
Entrez Gene ID: (Human) 4223, (Rat) 29279, (Mouse) 17286
Molecular Function: homeodomain transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support