Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MEHPLFGCLR SPHATAQGLH PFSQSSLALH GRSDHMSYPE LSTSSSSCII AGYPNEEGMF ASQHHRGHHH HHHHHHHHHQ QQQHQALQTN WHLPQMSSPP SAARHSLCLQ PDSGGPPELG SSPPVLCSNS SSLGSSTPTG AACAPGDYGR QALSPAEAEK RSGGKRKSDS SDSQEGNYKS EVNSKPRKER TAFTKEQIRE LEAEFAHHNY LTRLRRYEIA VNLDLTERQV KVWFQNRRMK WKRVKGGQQG AAAREKELVN VKKGTLLPSE LSGIGAATLQ QTGDSIANED SHDSDHSSEH AHL
Mesodermal transcription factor that plays a key role in somitogenesis and is required for sclerotome development. Activates expression of CDKN1A and CDKN2A in endothelial cells, acting as a regulator of vascular cell proliferation. While it activates CDKN1A in a DNA-dependent manner, it activates CDKN2A in a DNA-independent manner. May have a regulatory role when quiescent vascular smooth muscle cells reenter the cell cycle.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Growth arrest-specific homeobox; Homeobox protein MOX-2; mesenchyme homeo box 2 (growth arrest-specific homeo box); Mesenchyme homeobox 2
Gene Aliases: AI528662; GAX; MEOX2; Mox-2; MOX2
UniProt ID: (Human) P50222, (Rat) P39020, (Mouse) P32443
Entrez Gene ID: (Human) 4223, (Rat) 29279, (Mouse) 17286
Molecular Function: homeodomain transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support