Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: RRCRQRGDAG SPRGCPLCPD DCPAQMQTRT QAEGQGQGRG LGLVSAVRAG ASWGSPPACP GARAAPEPRD PPALFAEAAC PEGQAMEPSP PSPLLPGDVL ALLETWMPLS PLEWLPEEPK; Positive Samples: A-549, HeLa, THP-1, Mouse heart, Rat heart; Cellular Location: Nucleus
Mesp1 (mesoderm posterior 1) is a basic helix-loop-helix-type transcription factor, which along with its related proteins, has an essential role in both segmentation and rostro-caudal patterning of somites in the anterior pre-somitic mesoderm. Mesp1 is expressed in many of the precursors of the cardiovascular system and is known to play a role in the process of cardiac morphogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bHLHc5; Class C basic helix-loop-helix protein 5; mesoderm posterior 1 homolog; mesoderm posterior basic helix-loop-helix transcription factor 1; Mesoderm posterior protein 1; MGC10676
Gene Aliases: BHLHC5; MESP1
UniProt ID: (Human) Q9BRJ9, (Mouse) P97309
Entrez Gene ID: (Human) 55897, (Rat) 308766, (Mouse) 17292
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support