Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MAQPLCPPLS ESWMLSAAWG PTRRPPPSDK DCGRSLVSSP DSWGSTPADS PVASPARPGT LRD
Mesp1 (mesoderm posterior 1) is a basic helix-loop-helix-type transcription factor, which along with its related proteins, has an essential role in both segmentation and rostro-caudal patterning of somites in the anterior pre-somitic mesoderm. Mesp1 is expressed in many of the precursors of the cardiovascular system and is known to play a role in the process of cardiac morphogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bHLHc5; Class C basic helix-loop-helix protein 5; mesoderm posterior 1 homolog; mesoderm posterior basic helix-loop-helix transcription factor 1; Mesoderm posterior protein 1; MGC10676
Gene Aliases: BHLHC5; MESP1
UniProt ID: (Human) Q9BRJ9
Entrez Gene ID: (Human) 55897
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support