Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%).
This antibody has specificity for Human MAP1D/Methionine Aminopeptidase 1D.
The N-terminal methionine excision pathway is an essential process in which the N-terminal methionine is removed from many proteins, thus facilitating subsequent protein modification. In mitochondria, enzymes that catalyze this reaction are celled methionine aminopeptidases (MetAps, or MAPs; EC 3.4.11.18).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CDS of metAP-3 within PCR fragment; MAP 1D; metAP 1D; Methionine aminopeptidase 1D, mitochondrial; Methionyl aminopeptidase type 1D, mitochondrial; Peptidase M 1D
Gene Aliases: MAP 1D; MAP1D; MetAP 1D; METAP1D; Metap1l
UniProt ID: (Human) Q6UB28
Entrez Gene ID: (Human) 254042
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support