Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
METT10D is a probable methyltransferase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: methyltransferase 10 domain containing; Methyltransferase 10 domain-containing protein; Methyltransferase-like protein 16; N6-adenosine-methyltransferase METTL16; putative methyltransferase METT10D; RNA N6-adenosine-methyltransferase METTL16; U6 small nuclear RNA (adenine-(43)-N(6))-methyltransferase
Gene Aliases: METT10D; METTL16
UniProt ID: (Human) Q86W50
Entrez Gene ID: (Human) 79066
Molecular Function: methyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support