Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 1
Store as concentrated solution.
Centrifuge briefly prior to opening vial.
This gene is similar in sequence to the S. cerevisiae YDL201w gene. The gene product contains a conserved S-adenosylmethionine-binding motif and is inactivated by phosphorylation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: D1075-like gene product; Methyltransferase-like protein 1; METTL1; miRNA (guanine-N(7)-)-methyltransferase; mRNA (guanine-N(7)-)-methyltransferase; tRNA (guanine(46)-N(7))-methyltransferase; tRNA (guanine-N(7)-)-methyltransferase; tRNA(m7G46)-methyltransferase
Gene Aliases: C12orf1; METTL1; TRM8; TRMT8; YDL201w
UniProt ID: (Human) Q9UBP6
Entrez Gene ID: (Human) 4234
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support