Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
KIAA1627 is a probable methyltransferase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hMETTL14; Methyltransferase-like protein 14; N6-adenosine-methyltransferase non-catalytic subunit; N6-adenosine-methyltransferase subunit METTL14; snoRNA MBII-84
Gene Aliases: G430022H21Rik; KIAA1627; METTL14; mKIAA1627
UniProt ID: (Human) Q9HCE5, (Mouse) Q3UIK4
Entrez Gene ID: (Human) 57721, (Mouse) 210529
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support