Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MASLQRKGL QARILTSEEE EKLKRDQTLV SDFKQQKLEQ EAQKNWDLFY KRNSTNFFKD RHWTTREFEE LRSCREFEDQ KLTMLEAGCG VGNCLFPLLE EDPNIFAYAC DFSPRAIEYV KQNPLYDTER CKVFQCDLTK DDLLDHVPPE SVDVVMLIFV LSAVHPDKMH LVLQNIYKVL KPGKSVLFRD YGLYDHAMLR FKASSKLGEN FYVRQDGTRS YFFTDDFLAQ LFMDTGYEEV VNEYVFRETV N (1-250 aa encoded by BC022400)
S-adenosyl-L-methionine-dependent methyltransferase that mediates N(3)-methylcytidine modification of residue 32 of the tRNA anticodon loop of tRNA(Ser). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hMETTL6; Methyltransferase-like protein 6; tRNA N(3)-methylcytidine methyltransferase METTL6
Gene Aliases: 1600013P15Rik; AI467521; AU020711; METTL6
UniProt ID: (Human) Q8TCB7, (Mouse) Q8BVH9, (Rat) Q6AXU8
Entrez Gene ID: (Human) 131965, (Mouse) 67011, (Rat) 290564
Molecular Function: methyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support