Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Plays a role in mitochondrial and peroxisomal fission. Promotes the recruitment and association of the fission mediator dynamin-related protein 1 (DNM1L) to the mitochondrial surface. May be involved in regulation of synaptic vesicle membrane dynamics by recruitment of DNM1L to clathrin-containing vesicles. [Uniprot]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Mitochondrial fission factor
Gene Aliases: 5230400G24Rik; AD030; AD033; AI314724; C2orf33; GL004; MFF; RGD1310230
UniProt ID: (Human) Q9GZY8, (Rat) Q4KM98, (Mouse) Q6PCP5
Entrez Gene ID: (Human) 56947, (Rat) 301563, (Mouse) 75734
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support