Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VIPILVIAC DRSTVRRCLD KLLHYRPSAE LFPIIVSQDC GHEETAQAIA SYGSAVTHIR QPDLSSIAVP PDHRKFQGYY KIARHYRWAL GQVFRQFRFP AAVVVEDDLE VAPDFFEYFR ATYPLLKADP SLWCVSAWND NGKEQMVDAS RPELLYRTDF FPGLGWLLLA ELWAELEPKW PKAFWDDWMR RPEQRQGRAC IRPEISRTMT FGRKGVSHGQ FFDQHLKFIK LNQQFVHFTQ LDLSYLQREA YDRDFLARVY GAPQLQVEKV RTNDRKELGE VRVQYTGRDS FKAFAKALGV MDDLKSGVPR AGYRGIVTFQ FRGRRVHLAP PPTWEGYDPS WN (105-445 aa encoded by BC003575)
There are believed to be over 100 different glycosyltransferases involved in the synthesis of protein-bound and lipid-bound oligosaccharides. UDP-N-acetylglucosamine:alpha-3-D-mannoside beta-1, 2-N-acetylglucosaminyltransferase I is a medial-Golgi enzyme essential for the synthesis of hybrid and complex N-glycans. The protein, encoded by a single exon, shows typical features of a type II transmembrane protein. The protein is believed to be essential for normal embryogenesis. Several variants encoding the same protein have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha-1,3-mannosyl-glycoprotein 2-beta-N-acetylglucosaminyltransferase; glcNAc-T I; GNT-I; mannoside acetylglucosaminyltransferase 1; MGAT1 gene for N-acetylglucosaminyltransferase; N-acetylglucosaminyltransferase I; N-glycosyl-oligosaccharide-glycoprotein N-acetylglucosaminyltransferase I; OTTHUMP00000161546
Gene Aliases: GGNT1; GLCNAC-TI; GLCT1; GLYT1; GNT-1; GNT-I; Gnt1; GnTI; MGAT; Mgat-1; MGAT1
UniProt ID: (Human) P26572, (Rat) Q09325, (Mouse) P27808
Entrez Gene ID: (Human) 4245, (Rat) 81519, (Mouse) 17308
Molecular Function:
protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support