Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: PFSIQLYPNE FPGSDPRDCP RDLPKNAALK LGCINAEYPD SFGHYREAKF SQTKHHWWWK LHFVWERVKI LRDYAGLILF LEEDHYLAPD FYHVFKKMWK LKQQECPECD VLSLGTYSAS RSFYGMADKV DVKTWKSTEH NMGLALTRNA YQKLIECTDT FCTYDDYNWD WTLQYLTVSC LPKFWKVLVP QIPRIFHAGD CGMHHKKTCR PSTQSAQIES LLNNNKQYMF PETLTISEKF TVVAISPPRK NGGWGDIRDH ELCKSYRRLQ; Positive Samples: K-562, Mouse kidney, Rat kidney; Cellular Location: Golgi apparatus membrane, Single-pass type II membrane protein
The product of this gene is a Golgi enzyme catalyzing an essential step in the conversion of oligomannose to complex N-glycans. The enzyme has the typical glycosyltransferase domains: a short N-terminal cytoplasmic domain, a hydrophobic non-cleavable signal-anchor domain, and a C-terminal catalytic domain. Mutations in this gene may lead to carbohydrate-deficient glycoprotein syndrome, type II. The coding region of this gene is intronless. Transcript variants with a spliced 5' UTR may exist, but their biological validity has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha-1,6-mannosyl-glycoprotein 2-beta-N-acetylglucosaminyltransferase; Beta-1,2-N-acetylglucosaminyltransferase II; beta-12-N-acetylglucosaminyltransferase II; GlcNAc-T II; GNT-II; Mannoside acetylglucosaminyltransferase 2; N-glycosyl-oligosaccharide-glycoprotein N-acetylglucosaminyltransferase II; UDP-N-acetylglucosamine:alpha-6-D-mannoside beta-1,2-N-acetylglucosaminyltransferase II
Gene Aliases: AA407964; CDG2A; CDGS2; GLCNACTII; GNT-II; GNT2; MGAT2
UniProt ID: (Human) Q10469, (Mouse) Q921V5, (Rat) Q09326
Entrez Gene ID: (Human) 4247, (Mouse) 217664, (Rat) 94273
Molecular Function: glycosyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support