Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: LAEPGQSCKQ VCQESQLICE PSFFQHLNKD KDMLKYKVTC QSSELAKDIL VPSFDPKNKH CVFQGDLLLF SCAGAHPRHQ RVCPCRDFIK GQVALCKD
Catalyzes the addition of N-acetylglucosamine in beta 1- 6 linkage to the alpha-linked mannose of biantennary N-linked oligosaccharides. It is one of the most important enzymes involved in the regulation of the biosynthesis of glycoprotein oligosaccharides.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha-1,6-mannosylglycoprotein 6-beta-N-acetylglucosaminyltransferase A; alpha-mannoside beta-1,6-N-acetylglucosaminyltransferase; Alpha-mannoside beta-1,6-N-acetylglucosaminyltransferase V; EC 2.4.1.155; GlcNAc-T V; GNT-V; GNT-VGNT-VA; Mannoside acetylglucosaminyltransferase 5; N-acetylglucosaminyl-transferase V
Gene Aliases: GGNT5; GNT-V; GNT-VA; MGAT5
UniProt ID: (Human) Q09328
Entrez Gene ID: (Human) 4249
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support