Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 21
MGC12965 functions in DNA binding.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC12965
Entrez Gene ID: (Human) 100287896
Molecular Function: viral or transposable element protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support