Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MGC12965 functions in DNA binding.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC12965
Entrez Gene ID: (Human) 100287896
Molecular Function: viral or transposable element protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support