Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSQPRTPEQA LDTPGDCPPG RRDEDAGEGI QCSQRMLSFS DALLSIIATV MILPVTHTEI SPEQQFDRSV QRLLATRIAV YLMTFLIVTV AWAAHTRLFQ VVGKTDDTLA LLNLACMMTI TFLPYTFSLM VTFPDVPLGI FLFCVCVIAI GVVQALIVGY AFHFPHLLSP QIQRSAHRAL YRRHVLGIVL QGPALCFAAA IFSLFFVPLS YLLMVTVILL PYVSKVTGWC RDRLLGHREP SAHPVEVFSF DLHEPLSKER VEAFSDGVYA IVATLLILDI CEDNVPDPKD VKERFSGSLV AALSATGPRF LAYFGSFATV GLLWFAHHSL FLHVRKATRA MGLLNTLSLA FVGGLPLAYQ QTSAFARQPR DELERVRVSC TIIFLASIFQ LAMWTTALLH QAETLQPSVW FGGREHVLMF AKLALYPCAS LLAFASTCLL SRFSVGIFHL MQIAVPCAFL LLRLLVGLAL ATLRVLRGLA RPEHPPPAPT GQDDPQSQLL PAPC
TMEM175 gene ontology annotations related to this gene include potassium channel activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Endosomal/lysosomal proton channel TMEM175; hTMEM175; Potassium channel TMEM175; Transmembrane protein 175
Gene Aliases: hTMEM175; TMEM175
UniProt ID: (Human) Q9BSA9
Entrez Gene ID: (Human) 84286
Molecular Function:
ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support