Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MAGNSILLAA VSILSACQQS YFALQVGKAR LKYKVTPPAV TGSPEFERVF RAQQNCVEFY PIFIITLWMA GWYFNQVFAT CLGLVYIYGR HLYFWGYSEA AKKRITGFRL SLGILALLTL LGALGIANSF LDEYLDLNIA KKLRRQF
MGST2 (Microsomal glutathione S-transferase 2) is a 147 amino acid protein encoded by the human gene MGST2. MGST2 can catalyze the production of LTC4 (leukotriene C4) from LTA4 (leukotriene A4) and reduced glutathione. It can also catalyze the conjugation of 1-chloro-2,4-dinitrobenzene with reduced glutathione. MSGT2 is a multi-pass membrane protein found as a homodimer. MGST2 belongs to the MAPEG family and is expressed in liver, spleen, skeletal muscle, heart, adrenals, pancreas, prostate, testis, fetal liver, and fetal spleen. It has very low expression in lung, brain, placenta and bone marrow. Also, MGST2 is the only GST expressed in human umbilical vein endothelial cells (HUVECs).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ27438; Glutathione peroxidase MGST2; Leukotriene C4 synthase MGST2; MGC14097; Microsomal glutathione S-transferase 2; Microsomal glutathione S-transferase II; Microsomal GST-2; Microsomal GST-II
Gene Aliases: GST2; MGST-II; MGST2
UniProt ID: (Human) Q99735
Entrez Gene ID: (Human) 4258
Molecular Function:
transferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support